(R)-(+)-2-Phenyl-1-propanol, 98+%
Thermo Scientific Chemicals

(R)-(+)-2-Phenyl-1-propanol, 98+%

Have Questions?
Catalog NumberQuantity
L13999.03
also known as L13999-03
1 g
Catalog number L13999.03
also known as L13999-03
Price (EUR)
194,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
194,00
Each
Chemical Identifiers
CAS19141-40-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Optical Rotation+17.0° ± 1.0° (neat)
Refractive Index1.5250-1.5290 @ 20°C
For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For synthesis of optically active products