3-Aminocoumarin, 97%
3-Aminocoumarin, 97%
3-Aminocoumarin, 97%
Thermo Scientific Chemicals

3-Aminocoumarin, 97%

CAS: 1635-31-0 | C9H7NO2 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2 g
Catalog number L14031.04
also known as L14031-04
Price (USD)
235.65
Special offer
Online exclusive
Ends: 30-Jun-2026
277.00
Save 41.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
235.65
Special offer
Online exclusive
Ends: 30-Jun-2026
277.00
Save 41.35 (15%)
Each
Chemical Identifiers
CAS1635-31-0
IUPAC Name3-amino-2H-chromen-2-one
Molecular FormulaC9H7NO2
InChI KeyQWZHDKGQKYEBKK-UHFFFAOYSA-N
SMILESNC1=CC2=CC=CC=C2OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥96.0%
Appearance (Color)White to cream or yellow
Melting Point (clear melt)132-141.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only