Dimethoxymethylvinylsilane, 97%
Dimethoxymethylvinylsilane, 97%
Dimethoxymethylvinylsilane, 97%
Thermo Scientific Chemicals

Dimethoxymethylvinylsilane, 97%

CAS: 16753-62-1 | C5H12O2Si | 132.234 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L14035.22
also known as L14035-22
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
166.65
Online Exclusive
185.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS16753-62-1
IUPAC Nameethenyldimethoxymethylsilane
Molecular FormulaC5H12O2Si
InChI KeyZLNAFSPCNATQPQ-UHFFFAOYSA-N
SMILESCO[Si](C)(OC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3925-1.3975 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow to pale green
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only