Appearance (Color)Colorless to white to yellow
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Assay (Silylated GC)≥97.5%
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation-10.7 ± 0.5? (C=10 in water)
View more