Thermo Scientific Chemicals

Tri-n-butyl phosphite, 94%, Thermo Scientific Chemicals

Catalog number: L14307.AE
100 mL, Each
Thermo Scientific Chemicals

Tri-n-butyl phosphite, 94%, Thermo Scientific Chemicals

Catalog number: L14307.AE
100 mL, Each
Quantity
Catalog number: L14307.AE
also known as L14307-AE
Price (USD)

Chemical Identifiers

CAS
102-85-2
Impurity content
Dibutyl hydrogen phosphite (DBHP): ≤4.0%
Appearance (Color)
Colorless
Assay from Supplier's CofA
≥93.0%
Form
Liquid

Description

Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tributyl phosphite may be employed as model lubricant additive and its surface chemistry on Fe3O4 in ultrahigh vacuum is investigated. It is used as lubricants, preservatives, heat stabilizers of intensifier.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Store under inert gas. Store away from oxidizing agent, air, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text