trans,trans-Farnesol, 97%
trans,trans-Farnesol, 97%
trans,trans-Farnesol, 97%
trans,trans-Farnesol, 97%
Thermo Scientific Chemicals

trans,trans-Farnesol, 97%

CAS: 106-28-5 | C15H26O | 222.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L14348.03
also known as L14348-03
Price (EUR)
44,90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
44,90
Each
Add to cart
Chemical Identifiers
CAS106-28-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear, colorless to pale yellow
Refractive Index1.4875 - 1.4925
Assay from Supplier's CofA≥96.0%
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Terpene building block.
  2. A convenient 2-step alternative to the Gabriel or azide displacement methods for the synthesis of allylic amines has been reported using a Mitsunobu reagent (from Diisopropyl azodicarboxyl ate, L10386) in combination with phthalimide, followed by mild deprotection with methylamine. This technique avoids the allylic rearrangements and other side reactions associated with the older routes: Synthesis, 756 (1995):
  3. Espinosa, R. R.; Inchingolo, R.; Alencar, S. M.; Rodriguez-Estrada, M. T.; Castro, I. A. Antioxidant activity of phenolic compounds added to a functional emulsion containing μ-3 fatty acids and plant sterol esters. Food Chem. 2015, 182, 95-104.
  4. Nikolić, B.; Ristić, M.; Bojović, S.; Krivošej, Z.; Matevski, V.; Marin, P. D. Population Variability of Essential Oils of Pinus heldreichii from the Scardo-Pindic Mountains Ošljak and Galičica. Chem. Biodivers. 2015, 12 (2), 295-308.