11-Bromoundecanol, 97%
11-Bromoundecanol, 97%
11-Bromoundecanol, 97%
Thermo Scientific Chemicals

11-Bromoundecanol, 97%

CAS: 1611-56-9 | C11H23BrO | 251.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 L14448.14
亦稱為 L14448-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS1611-56-9
IUPAC Name11-bromoundecan-1-ol
Molecular FormulaC11H23BrO
InChI KeyXFGANBYCJWQYBI-UHFFFAOYSA-N
SMILESOCCCCCCCCCCCBr
檢視更多
規格Specification Sheet規格表
Melting Point41.5-50.5?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Frey BL, et al. Vibrational spectroscopic studies of the attachment chemistry for zirconium phosphonate multilayers at gold and germanium surfaces. Langmuir 1993, 9 (7), 1815-20.
  2. Feng Yan; and John Texter. Surfactant ionic liquid-based microemulsions for polymerization.. Chemical Communications. 2006, 25 (1-3), 2696-2698.