4-tert-Butylcalix[6]arene, 96%
4-tert-Butylcalix[6]arene, 96%
4-tert-Butylcalix[6]arene, 96%
4-tert-Butylcalix[6]arene, 96%
Thermo Scientific Chemicals

4-tert-Butylcalix[6]arene, 96%

CAS: 78092-53-2 | C66H84O6 | 973.392 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L14473.03
also known as L14473-03
Price (EUR)
27,70
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
27,70
Each
Add to cart
Chemical Identifiers
CAS78092-53-2
IUPAC Name5,11,17,23,29,35-hexa-tert-butylheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Molecular FormulaC66H84O6
InChI KeyUOEYZAXKBKAKRO-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(C2)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥95.0%
FormPowder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only