(S)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
(S)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
(S)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene
Thermo Scientific Chemicals

(S)-2-Methyl-CBS-oxazaborolidine, 1M soln. in toluene

CAS: 112022-81-8 | C18H20BNO | 277.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L14583.AA
also known as L14583-AA
Price (USD)
77.65
Online Exclusive
86.10
Save 8.45 (10%)
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.10
Save 8.45 (10%)
Each
Chemical Identifiers
CAS112022-81-8
IUPAC Name(3aS)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular FormulaC18H20BNO
InChI KeyVMKAFJQFKBASMU-KRWDZBQOSA-N
SMILES[H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to yellow
FormLiquid
Assay0.95-1.10M
Assay28.5-33.0% (theory 30.0%)
Assay (unspecified)0.95-1.10M
View more
(S)-2-Methyl-CBS-oxazaborolidine is used as an oxazaborolidine catalyst. It is also employed in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2-symmetrical ferrocenyl diols and propargyl alcohols. It is an anhydrous catalyst for CBS reduction reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-Methyl-CBS-oxazaborolidine is used as an oxazaborolidine catalyst. It is also employed in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2-symmetrical ferrocenyl diols and propargyl alcohols. It is an anhydrous catalyst for ′CBS′ reduction reaction.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Panayiotis A Procopiou.; Gillian E Morton.; Martin Todd.; Graham Webb. Enantioselective synthesis of (S)-salmeterol via asymmetric reduction of azidoketone by Pichia angusta. Tetrahedron: Asymmetry. 2001, 12 (14), 2005-2008.
  2. Toshiyasu Takemoto.; Katsuyoshi Nakajima.; Yukiko Iio, Masakazu Tamura.; Takahide Nishi. Asymmetric synthesis of enantiomerically pure spiro[((2S)-hydroxy)indane-1,4'-piperidine]. Tetrahedron: Asymmetry. 1999, 10 (9), 1787-1793.
  3. Anhydrous catalyst for CBS reduction reaction. For examples of use in asymmetric reduction of ketones, see: Org. Synth. Coll., 9, 362, 676 (1998). For further information, see (S)-2-Methyl-CBS-oxazaborolidine monohydrate, L09219.