Thermo Scientific Chemicals

Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L14619.AE
100 mL, Each
Thermo Scientific Chemicals

Isoprene, 99%, stab. with ca 0.02% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L14619.AE
100 mL, Each
Quantity
Catalog number: L14619.AE
also known as L14619-AE
Price (USD)
Price: 36.40
Your price:
Quantity
-

Chemical Identifiers

CAS
78-79-5
IUPAC Name
2-methylbuta-1,3-diene
Molecular Formula
C5H8
InChI Key
RRHGJUQNOFWUDK-UHFFFAOYSA-N
SMILES
CC(=C)C=C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5% (non-U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries
Refractive Index
1.4205-1.4235 @ 20°C (non-U.S. sourced material)
Identification (FTIR)
Conforms

Description

Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoprene is used in to prepare synthetic version of natural rubber by polymerization using Ziegler catalysts. It is a basic building block for terpenes especially carotenes, terpenoids, retinol, vitamin-A and tocopherol (Vitamin-E). It finds applications in medical equipments, toys, baby bottle nipples, shoe soles, elastic films and threads for golf balls. Copolymer prepares in association with styrene is used in pressure sensitive adhesives.

Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone and benzene. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, ammonia, alkali metals, alcohols and oxygen.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text