2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals
2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals
2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L14664.03
also known as L14664-03
1 g
Catalog number L14664.03
also known as L14664-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein1749961
CAS813-72-9
Chemical Name or Material2,2-Dimethylhexanoic acid
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Density0.913
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only