Bisphenol A diglycidyl ether resin
Bisphenol A diglycidyl ether resin
Thermo Scientific Chemicals

Bisphenol A diglycidyl ether resin

CAS: 1675-54-3 | C21H24O4 | 340.419 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L14817.14
also known as L14817-14
Price (EUR)
28,50
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
28,50
Each
Add to cart
Chemical Identifiers
CAS1675-54-3
IUPAC Name2-{[4-(2-{4-[(oxiran-2-yl)methoxy]phenyl}propan-2-yl)phenoxy]methyl}oxirane
Molecular FormulaC21H24O4
InChI KeyLCFVJGUPQDGYKZ-UHFFFAOYNA-N
SMILESCC(C)(C1=CC=C(OCC2CO2)C=C1)C1=CC=C(OCC2CO2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
FormLiquid or viscous liquid
Refractive Index1.500-1.640 at 20°C
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bisphenol A diglycidyl ether is used to inhibit the peroxisome proliferator-activated receptor (PPARγ) and adipocyte differentiation and it induce apoptosis in tumor cells.

Solubility
Soluble in 100% ethanol, dimethyl sulfoxide (100 mM), dimethyl formamide, chloroform, methanol, and ethanol (50 mM). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Neus Casajuana.; Sílvia Lacorte. New Methodology for the Determination of Phthalate Esters, Bisphenol A, Bisphenol A Diglycidyl Ether, and Nonylphenol in Commercial Whole Milk Samples . J. Agric. Food Chem. 2004, 52 (12), 3702-3707.
  2. P. Paseiro Losada.; J. Simal Lozano.; S. Paz Abuín.; P. López Mahía.; J. Simal Gándara. Kinetics of the hydrolysis of bisphenol A diglycidyl ether (BADGE) in water-based food simulants. Fresenius' Journal of Analytical Chemistry. 1993, 345 (7), 527-532.