Cyclooctane, 99+%
Cyclooctane, 99+%
Cyclooctane, 99+%
Thermo Scientific Chemicals

Cyclooctane, 99+%

CAS: 292-64-8 | C8H16 | 112.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Catalog number L15000.AK
also known as L15000-AK
Price (EUR)
161,00
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
161,00
Each
Add to cart
Chemical Identifiers
CAS292-64-8
IUPAC Namecyclooctane
Molecular FormulaC8H16
InChI KeyWJTCGQSWYFHTAC-UHFFFAOYSA-N
SMILESC1CCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥99.0%
Refractive Index1.4570-1.4600 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Gerald K. Cook.; James M. Mayer. C-H Bond Activation by Metal Oxo Species: Chromyl Chloride Oxidations of Cyclooctane, Isobutane, and Toluene. J. Am. Chem. Soc. 1995, 117 (27), 7139-7156.
  2. F. A. L. Anet.; V. J. Basus. Detection of a crown family conformation in cyclooctane by proton and carbon-13 nuclear magnetic resonance. J. Am. Chem. Soc. 1973, 95 (13), 4424-4426.