Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle
Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle
Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle

CAS: 4039-32-1 | C6H18LiNSi2 | 167.327 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L15012.9Z
also known as L15012-9Z
Price (TWD)
-
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS4039-32-1
IUPAC Namelithium(1+) bis(trimethylsilyl)azanide
Molecular FormulaC6H18LiNSi2
InChI KeyYNESATAKKCNGOF-UHFFFAOYSA-N
SMILES[Li+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear or slightly hazy, yellow to brown
FormLiquid
Assay (unspecified)18-22% active base as LHMDS
Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Bert Y. Kimura.; Theodore L. Brown. Solvent effects on the aggregation of lithium bis(trimethylsilyl)amide. Journal of Organometallic Chemistry. 1971, 26 (1), 57-67.
  2. Sunwoo Lee.; Morten Jørgensen.; John F. Hartwig. Palladium-Catalyzed Synthesis of Arylamines from Aryl Halides and Lithium Bis(trimethylsilyl)amide as an Ammonia Equivalent. Org. Lett. 2001, 3 (17), 2729-2732.
  3. Supplied in septum cap bottle.
  4. Versatile non-nucleophilic strong base. For examples of use in the lithiation of ethyl acetate, see: Org. Synth. Coll., 6, 598 (1988), and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones: Org. Synth. Coll., 9, 442 (1998).