Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L15164.14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS37342-97-5
IUPAC Nameλ²-zirconium(2+) bis(cyclopenta-2,4-dien-1-ide) chloride hydride
Molecular FormulaC10H11ClZr
InChI KeyVPQMECJLNPUNGN-UHFFFAOYSA-M
SMILES[H-].[Cl-].[Zr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Elemental Analysis(Carbon) : 45.65-47.51%
Proton NMRConforms to structure
Elemental Analysis(Hydrogen) : 4.21-4.39%
Impurity content(Dihydride) : ≤5.0%
View more
Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture, light and air. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jared T Spletstoser.; Jonathan M White.; Ashok Rao Tunoori.; Gunda I Georg. Mild and selective hydrozirconation of amides to aldehydes using Cp2Zr(H)Cl: scope and mechanistic insight.. Journal of the American Chemical Society. 2007, 129 (11), 3408-3419.
  2. Thomas E. Cole.; Ramona. Quintanilla.; Stephan. Rodewald. Migration of 1-alkenyl groups from zirconium to boron compounds. Organometallics. 1991, 10 (10), 3777-3781.