Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals

Catalog number: L15178.03
1 g, Each
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid dianhydride, 95%, Thermo Scientific Chemicals

Catalog number: L15178.03
1 g, Each
Quantity
Catalog number: L15178.03
also known as L15178-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
23911-26-4
IUPAC Name
2-{bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino}acetic acid
Molecular Formula
C14H19N3O8
InChI Key
RAZLJUXJEOEYAM-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCN1CC(=O)OC(=O)C1)CCN1CC(=O)OC(=O)C1
Form
Powder
Elemental Analysis
Hydrogen, H: 5.04 - 5.68% (Theory 5.36%)
Elemental Analysis
Nitrogen, N: 11.05 - 12.46% (Theory 11.77%)
Proton NMR
Conforms to structure
Elemental Analysis
Carbon, C: 44.24 - 49.88% (Theory 47.06%)

Description

Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid dianhydride is a reagent used for introducing a strong chelator. It is employed in the synthesis of macrocyclic ligands and lanthanide complexes that are useful as MRI enhancers.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is-20°C. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text