Amberlite™ IRC120 Na ion exchange resin
Amberlite™ IRC120 Na ion exchange resin
Amberlite™ IRC120 Na ion exchange resin
Thermo Scientific Chemicals

Amberlite™ IRC120 Na ion exchange resin

CAS: 78922-04-0 | C13H10ClNO4S | 311.736 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number L15199.22
also known as L15199-22
Price (EUR)
29,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
29,10
Each
Chemical Identifiers
CAS78922-04-0
IUPAC Name3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular FormulaC13H10ClNO4S
InChI KeyAPBOVLPLJFJSRI-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMax reversible swelling Na -> H ca. 10%
Exchange capacity≥2.00 eq/L (Na+ form)
Particle Size<300 µm : ≤2.0%
Appearance (Color)Pale brown to brown
MatrixStyrene divinylbenzene copolymer
View more
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.

Solubility
Insoluble in water.

Notes
Water softening, dealkalinization, deionization
RUO – Research Use Only

General References:

  1. José L. Valverde.; Antonio de Lucas.; Marcela González.; Juan F. Rodríguez. Ion-Exchange Equilibria of Cu2+, Cd2+, Zn2+, and Na+ Ions on the Cationic Exchanger Amberlite IR-120. J. Chem. Eng. Data. 2001, 46 1404-1409.
  2. Juan Francisco Rodríguez.; Antonio De Lucas.; Jose R. Leal.; Jose Luis Valverde. Determination of intraparticle diffusivities of Na+/K+ in water and water/alcohol mixed solvents on a strong acid cation exchanger. Ind. Eng. Chem. Res. 2002, 41 3019-3027.