L-Pipecolinic acid, 99%
L-Pipecolinic acid, 99%
L-Pipecolinic acid, 99%
L-Pipecolinic acid, 99%
Thermo Scientific Chemicals

L-Pipecolinic acid, 99%

CAS: 3105-95-1 | C6H11NO2 | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15373.03
also known as L15373-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS3105-95-1
IUPAC Namepiperidine-2-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyHXEACLLIILLPRG-UHFFFAOYNA-N
SMILESOC(=O)C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-26.6 ± 0.5? (c=1 in water)
Appearance (Color)White to cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormPowder
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Pipecolinic acid is involved in synaptic transmission in the central nervous system. It is also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R. I. Morrison. The isolation of l-pipecolinic acid from Trifolium repens. Biochem J. 1953, 53 (3), 474-478..
  2. Go Takei.; Takehiko Kitamori.; Haeng-Boo Kim. Photocatalytic redox-combined synthesis of l-pipecolinic acid with a titania-modified microchannel chip. Catalysis Communications. 2005, 6 (5), 357-360.