3',5'-Dimethylacetophenone, 97%
3',5'-Dimethylacetophenone, 97%
3',5'-Dimethylacetophenone, 97%
Thermo Scientific Chemicals

3',5'-Dimethylacetophenone, 97%

CAS: 5379-16-8 | C10H12O | 148.205 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L15480.03
also known as L15480-03
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Specifications
Beilstein2240987
CAS5379-16-8
Chemical Name or Material3',5'-Dimethylacetophenone
Density0.988
Flash Point93°C (199°F)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only