3-Isopropylbenzeneboronic acid, 99%
3-Isopropylbenzeneboronic acid, 99%
3-Isopropylbenzeneboronic acid, 99%
Thermo Scientific Chemicals

3-Isopropylbenzeneboronic acid, 99%

CAS: 216019-28-2 | C9H13BO2 | 164.01 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
產品號碼 L15530.03
亦稱為 L15530-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS216019-28-2
IUPAC Name[3-(propan-2-yl)phenyl]boronic acid
Molecular FormulaC9H13BO2
InChI KeyQSWLFBMVIGQONC-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC(=C1)B(O)O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5%
Assay (HPLC)≥98.5%
Proton NMRConforms to structure
FormPowder or granules
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only