Thermo Scientific Chemicals

Diethyl difluoromalonate, 97%, Thermo Scientific Chemicals

Catalog number: L16042.06
5 g, Each
Thermo Scientific Chemicals

Diethyl difluoromalonate, 97%, Thermo Scientific Chemicals

Catalog number: L16042.06
5 g, Each
Quantity
Catalog number: L16042.06
also known as L16042-06
Price (USD)
Price: 269.00
Online price: 232.65
Your price:
Quantity
-

Chemical Identifiers

CAS
680-65-9
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3765-1.3815 @ 20?C

Description

Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text