Carbon, activated, Norit ROW 0.8mm pellets, steam activated
Carbon, activated, Norit ROW 0.8mm pellets, steam activated
Carbon, activated, Norit ROW 0.8mm pellets, steam activated
Thermo Scientific Chemicals

Carbon, activated, Norit ROW 0.8mm pellets, steam activated

CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2000 g
Catalog number L16334.0P
also known as L16334-0P
Price (EUR)
193,00
Each
Quantity:
2000 g
Request bulk or custom format
Price (EUR)
193,00
Each
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Formgranules
Appearance (Color)Black
Iodine value≥ 1100
Water Content (Karl Fischer Titration)≤ 5.0%
Particle Size≤ 0.5% is < 0.60mm
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
RUO – Research Use Only

General References:

  1. Vasile V.N. Obreja. On the performance of supercapacitors with electrodes based on carbon nanotubes and carbon activated material—A review. Physica E: Low-dimensional Systems and Nanostructures. 2008, 40 (7), 2596-2605.
  2. Z. Hu.; M. P. Srinivasan.; Y. Ni. Preparation of Mesoporous High-Surface-Area Activated Carbon. Advanced Materials. 2000, 12 (1), 62-65.
  3. Extruded carbon with large pore volume and pore size distribution, suitable for decolorization, deodorisation and purification applications.