3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT
3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT
3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT
Thermo Scientific Chemicals

3-(Acryloyloxy)propyltrimethoxysilane, 94%, stab. with 100ppm BHT

CAS: 4369-14-6 | C9H18O5Si | 234.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16400.06
also known as L16400-06
Price (EUR)
99,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
99,60
Each
Chemical Identifiers
CAS4369-14-6
IUPAC Name3-(trimethoxysilyl)propyl prop-2-enoate
Molecular FormulaC9H18O5Si
InChI KeyKBQVDAIIQCXKPI-UHFFFAOYSA-N
SMILESCO[Si](CCCOC(=O)C=C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥92.5%
Identification (FTIR)Conforms
Refractive Index1.4255-1.4325 @ 20?C
3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Acryloyloxy)propyltrimethoxysilane is used to enhance the thermal conductivity of silicone rubber filled with hybrid fillers. It is involved in the synthesis of vinyl and acryalte modified silica nanoparticles by using vinyltrimethoxysilane. Further, it is used as a silane coupling agent involved in the modification of fluorescein isothiocyanate-mesoporous silica nanoparticles with carbon-carbon double bonds.

Solubility
Miscible with organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids and bases. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Gigot, A.; Sangermano, M.; Capozzi, L. C.; Dietliker, K. In-situ synthesis of organic-inorganic coatings via a photolatent base catalyzed Michael-addition reaction. Polymer 2015, 68, 195-201.
  2. Duffy, E.; Mitev, D. P.; Thickett, S. C.; Townsend, A. T.; Paull, B.; Nesterenko, P. N. Assessing the extent, stability, purity and properties of silanised detonation nanodiamond. Appl. Surf. Sci. 2015, 357, 397-406.