Thermo Scientific Chemicals

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog number: L16873.03
1 g, Each
Thermo Scientific Chemicals

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog number: L16873.03
1 g, Each
Quantity
Catalog number: L16873.03
also known as L16873-03
Price (USD)
Price: 51.50
Online price: 44.65
Your price:
Quantity
-

Chemical Identifiers

CAS
338-03-4
IUPAC Name
(3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Molecular Formula
C5H6F3O3
InChI Key
JCJGOWYCFRASTM-BYPYZUCNSA-M
SMILES
C[C@](O)(CC([O-])=O)C(F)(F)F
Appearance (Color)
White or clear colorless to pale yellow
Form
Fused solid or liquid as melt
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text