n-Decyltrimethoxysilane, 97%
n-Decyltrimethoxysilane, 97%
n-Decyltrimethoxysilane, 97%
Thermo Scientific Chemicals

n-Decyltrimethoxysilane, 97%

CAS: 5575-48-4 | C13H30O3Si | 262.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L16966.22
also known as L16966-22
Price (EUR)
150,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
150,00
Each
Chemical Identifiers
CAS5575-48-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4205-1.4255 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jing He.; Yan Xu.; Hui Ma.; Qiong Zhang.; David G. Evans.; Xue Duan. Effect of surface hydrophobicity/hydrophilicity of mesoporous supports on the activity of immobilized lipase. Journal of Colloid and Interface Science. 2006, 298 (2), 780-786.
  2. Marianna Kemell.; Elina Färm.; Markku Leskelä.; Mikko Ritala. Transparent superhydrophobic surfaces by self-assembly of hydrophobic monolayers on nanostructured surfaces. physica status solidi (a). 2006, 203 (6), 1453-1458.