Thermo Scientific Chemicals

3-Bromo-2-chloropyridine-5-sulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L17015.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-2-chloropyridine-5-sulfonyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: L17015.06
5 g, Each
Quantity
Catalog number: L17015.06
also known as L17015-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
216394-05-7
Appearance (Color)
White to cream to yellow
Melting Point
65-76°C
Form
Powder or crystalline powder
Assay (Titration ex Chloride)
≥92.0 to ≤108.0%

Description

3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-chloropyridine-5-sulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Reacts with water. Soluble in dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text