Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%
Thermo Scientific Chemicals

Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%

CAS: 72022-68-5 | C22H40N2S10Zn | 718.556 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L17046.03
also known as L17046-03
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
290.65
Online Exclusive
323.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS72022-68-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bright red
FormPowder
Elemental AnalysisCarbon: 35.30-38.24%
Elemental AnalysisNitrogen: 3.74-4.06%
Elemental AnalysisSulfur: 42.83-46.40%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only