Thermo Scientific Chemicals

Ethyl 4,4,4-trifluoro-2-methylacetoacetate, 96%, Thermo Scientific Chemicals

Catalog number: L17335.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 4,4,4-trifluoro-2-methylacetoacetate, 96%, Thermo Scientific Chemicals

Catalog number: L17335.06
5 g, Each
Quantity
Catalog number: L17335.06
also known as L17335-06
Price (USD)
Price: 103.00
Online price: 88.65
Your price:
Quantity
-

Chemical Identifiers

CAS
344-00-3
IUPAC Name
ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
Molecular Formula
C7H9F3O3
InChI Key
YLRGPBKEZVHOAW-UHFFFAOYNA-N
SMILES
CCOC(=O)C(C)C(=O)C(F)(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (Silylated GC)
≥95.0% (sum of enantiomers)
Identification (FTIR)
Conforms
Refractive Index
1.3665-1.3735 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text