Ethyl 4-acetyl-5-oxohexanoate, 98%
Thermo Scientific Chemicals

Ethyl 4-acetyl-5-oxohexanoate, 98%

CAS: 2832-10-2 | C11H21BrO2 | 265.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17496.06
also known as L17496-06
Price (EUR)
14,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
14,60
Each
Chemical Identifiers
CAS2832-10-2
IUPAC Nameethyl (4E)-4-acetyl-5-hydroxyhex-4-enoate
Molecular FormulaC10H16O4
InChI KeyCVPCFJRJUCNUJG-VQHVLOKHSA-N
SMILESCCOC(=O)CCC(=C(C)O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4530-1.4570 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only