4-Cyanobenzeneboronic acid, 98%
4-Cyanobenzeneboronic acid, 98%
4-Cyanobenzeneboronic acid, 98%
Thermo Scientific Chemicals

4-Cyanobenzeneboronic acid, 98%

CAS: 126747-14-6 | C7H6BNO2 | 146.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18007.03
also known as L18007-03
Price (USD)
55.65
Online Exclusive
62.00
Save 6.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
62.00
Save 6.35 (10%)
Each
Chemical Identifiers
CAS126747-14-6
IUPAC Name(4-cyanophenyl)boronic acid
Molecular FormulaC7H6BNO2
InChI KeyCEBAHYWORUOILU-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
Appearance (Color)White to pale cream to pale yellow
Mass SpecConsistent with structure
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only