Thermo Scientific Chemicals

Oxazole, 98+%, Thermo Scientific Chemicals

Catalog number: L18341.03
1 g, Each
Thermo Scientific Chemicals

Oxazole, 98+%, Thermo Scientific Chemicals

Catalog number: L18341.03
1 g, Each
Quantity
Catalog number: L18341.03
also known as L18341-03
Price (USD)
Price: 117.00
Online price: 99.65
Your price:
Quantity
-

Chemical Identifiers

CAS
288-42-6
IUPAC Name
1,3-oxazole
Molecular Formula
C3H3NO
InChI Key
ZCQWOFVYLHDMMC-UHFFFAOYSA-N
SMILES
O1C=CN=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4240-1.4280 @ 20°C

Description

Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text