Thermo Scientific Chemicals

Glyoxal dimethyl acetal, 60% aq. soln., Thermo Scientific Chemicals

Catalog number: L18490.0P
2000 g, Each
Thermo Scientific Chemicals

Glyoxal dimethyl acetal, 60% aq. soln., Thermo Scientific Chemicals

Catalog number: L18490.0P
2000 g, Each
Quantity
Catalog number: L18490.0P
also known as L18490-0P
Price (USD)
Price: 524.00
Online price: 445.65
Your price:
Quantity
-

Chemical Identifiers

CAS
51673-84-8
IUPAC Name
2,2-dimethoxyacetaldehyde
Molecular Formula
C4H8O3
InChI Key
OGFKTAMJLKHRAZ-UHFFFAOYSA-N
SMILES
COC(OC)C=O
Refractive Index
1.4105-1.4150 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (Titration)
≥58.0%
Identification (FTIR)
Conforms
Form
Liquid

Description

Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text