3,5-Dihydroxytoluene, 99%
3,5-Dihydroxytoluene, 99%
3,5-Dihydroxytoluene, 99%
Thermo Scientific Chemicals

3,5-Dihydroxytoluene, 99%

CAS: 504-15-4 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L18567.06
also known as L18567-06
Price (EUR)
51,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
51,40
Each
Chemical Identifiers
CAS504-15-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown or pale pink to pink
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point105-110°C
FormCrystalline powder or powder
Used in production of orcein

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only