Thermo Scientific Chemicals

Ambersep™ 900(OH), ion exchange resin, Thermo Scientific Chemicals

Catalog number: L19035.22
100 g, Each
Thermo Scientific Chemicals

Ambersep™ 900(OH), ion exchange resin, Thermo Scientific Chemicals

Catalog number: L19035.22
100 g, Each
Quantity
Catalog number: L19035.22
also known as L19035-22
Price (USD)
Price: 35.10
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9017-79-2
Moisture holding capacity
66.0-75.0 %
Particle Size
Harmonic Mean Diameter: 0.560-0.700mm
Form
Beads
Exchange capacity
(OH form): ≥0.80 meq/L
Particle Size
300-1180?m: ≥94.5%

Description

Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Specially sized for use in Ambersep resin systems and high-flow rate condensate polishing. It holds regenerable mixed bed applications such as the production of ultrapure water for semiconductor manufacturing.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text