Thermo Scientific Chemicals

Harmine, 98+%, Thermo Scientific Chemicals

Catalog number: L19068.03
1 g, Each
Thermo Scientific Chemicals

Harmine, 98+%, Thermo Scientific Chemicals

Catalog number: L19068.03
1 g, Each
Quantity
Catalog number: L19068.03
also known as L19068-03
Price (USD)
Price: 115.00
Online price: 98.65
Your price:
Quantity
-

Chemical Identifiers

CAS
442-51-3
Appearance (Color)
White to cream
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder

Description

Harmine, inhibits the Wnt signaling pathway in a cell-specific manner. Down-regulates inflammatory gene expression (TNF-α, IL-1β, iNOS). Reversibly inhibits MAO-A (monoamine oxidase A) but has no effect on MAO-B. Harmine is an inhibitor of Dyrk1A, Dyrk2 and Dyrk3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Harmine, inhibits the Wnt signaling pathway in a cell-specific manner. Down-regulates inflammatory gene expression (TNF-α, IL-1β, iNOS). Reversibly inhibits MAO-A (monoamine oxidase A) but has no effect on MAO-B. Harmine is an inhibitor of Dyrk1A, Dyrk2 and Dyrk3.

Solubility
Soluble in DMSO (100 mM), DMF (∼1.5 mg/ml), ethanol (∼1.5 mg/ml), and PBS (pH 7.2, ∼0.25 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text