2-Fluoro-5-nitrobenzaldehyde, 98%
2-Fluoro-5-nitrobenzaldehyde, 98%
2-Fluoro-5-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Fluoro-5-nitrobenzaldehyde, 98%

CAS: 27996-87-8 | C7H4FNO3 | 169.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19233.06
also known as L19233-06
Price (EUR)
121,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
121,00
Each
Chemical Identifiers
CAS27996-87-8
IUPAC Name2-fluoro-5-nitrobenzaldehyde
Molecular FormulaC7H4FNO3
InChI KeyVVXFDFQEIRGULC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to red
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or lumps or needles
Free acid (titration)≤1.5%
Melting Point (clear melt)56.0-62.0?C
2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. DJ Gale.; JFK Wilshire. The preparation of some polymethine astrazon dyes. Australian Journal of Chemistry. 197023 (5), 1063 - 1068.
  2. X Ouyang.; AS Kiselyov. Novel synthesis of dibenzo [b, g] 1, 5-oxazocines. Tetrahedron letters. 199940 (32), 5827-5830.