5-(3-Methylphenyl)-1H-tetrazole, 99%
5-(3-Methylphenyl)-1H-tetrazole, 99%
5-(3-Methylphenyl)-1H-tetrazole, 99%
Thermo Scientific Chemicals

5-(3-Methylphenyl)-1H-tetrazole, 99%

CAS: 3441-00-7 | C8H8N4 | 160.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19263.MD
also known as L19263-MD
Price (TWD)
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Beilstein131205
CAS3441-00-7
Chemical Name or Material5-(3-Methylphenyl)-1H-tetrazole
Formula Weight160.18
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only