5-Aminomethyl-2-chloropyridine, 97%
5-Aminomethyl-2-chloropyridine, 97%
5-Aminomethyl-2-chloropyridine, 97%
Thermo Scientific Chemicals

5-Aminomethyl-2-chloropyridine, 97%

CAS: 97004-04-1 | C6H7ClN2 | 142.586 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
產品號碼 L19283.06
亦稱為 L19283-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS97004-04-1
IUPAC Name1-(6-chloropyridin-3-yl)methanamine
Molecular FormulaC6H7ClN2
InChI KeyXPARFBOWIYMLMY-UHFFFAOYSA-N
SMILESNCC1=CN=C(Cl)C=C1
檢視更多
規格Specification Sheet規格表
Melting Point25-33?C (clear to cloudy melt)
FormPowder or Fused solid
Appearance (Color)White to yellow or pale cream to pale brown
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only