(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%
(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%
(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%
(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%
Thermo Scientific Chemicals

(S)-(-)-N-Benzyl-1-phenylethylamine, ChiPros∣r, 99%, ee 99+%

CAS: 17480-69-2 | C15H24N | 218.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19299.14
also known as L19299-14
Price (EUR)
115,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
115,00
Each
Add to cart
Chemical Identifiers
CAS17480-69-2
IUPAC Name(cyclohexylmethyl)[(1S)-1-phenylethyl]azanium
Molecular FormulaC15H24N
InChI KeyUHABCGJJMSQRRP-ZDUSSCGKSA-O
SMILESC[C@H]([NH2+]CC1CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5625-1.5655
Appearance (Color)Clear, colourless to pale yellow
Formliquid
Assay (GC)>98.5%
Optical Rotation-39? ?2? (neat)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only