1,3-Benzenedithiol, 97%
1,3-Benzenedithiol, 97%
1,3-Benzenedithiol, 97%
Thermo Scientific Chemicals

1,3-Benzenedithiol, 97%

CAS: 626-04-0 | C6H6S2 | 142.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19439.06
also known as L19439-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS626-04-0
IUPAC Namebenzene-1,3-dithiol
Molecular FormulaC6H6S2
InChI KeyZWOASCVFHSYHOB-UHFFFAOYSA-N
SMILESSC1=CC(S)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
Assay (GC)>96.0%
Refractive Index1.6615-1.6675 @ 20?C
FormLiquid
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong bases.
RUO – Research Use Only

General References:

  1. Huang, C.; Rudnev, A. V.; Hong, W.; Wandlowski, T. Break junction under electrochemical gating: testbed for single-molecule electronics. Chem. Soc. Rev. 2015, 44 (4), 889-901.
  2. Xu, F.; Gerlein, L. F.; Ma, X.; Haughn, C. R.; Doty, M. F.; Cloutier, S. G. Impact of Different Surface Ligands on the Optical Properties of PbS Quantum Dot Solids. Materials 2015, 8 (4), 1858-1870.