Thermo Scientific Chemicals

4-(Trifluoromethyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L19441.MD
250 mg, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: L19441.MD
250 mg, Each
Quantity
Catalog number: L19441.MD
also known as L19441-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
155849-49-3
IUPAC Name
4-(trifluoromethyl)piperidine hydrochloride
Molecular Formula
C6H11ClF3N
InChI Key
KMUKXPQVPQUCDB-UHFFFAOYSA-N
SMILES
Cl.FC(F)(F)C1CCNCC1
Appearance (Color)
White to yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text