3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Diethoxy-1-propylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
產品號碼Quantity
L19532.03
亦稱為 L19532-03
1 g
產品號碼 L19532.03
亦稱為 L19532-03
價格 (TWD)
-
申請報價
Quantity:
1 g
Request bulk or custom format
化學識別
CAS165904-27-8
IUPAC Name2-(3,3-diethoxypropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC13H27BO4
InChI KeyVEDSPUPKZZMMLL-UHFFFAOYSA-N
SMILESCCOC(CCB1OC(C)(C)C(C)(C)O1)OCC
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Refractive Index1.4260-1.4310 @ 20?C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only