3-Chloro-2,5,6-trifluoropyridine, 98+%
3-Chloro-2,5,6-trifluoropyridine, 98+%
3-Chloro-2,5,6-trifluoropyridine, 98+%
Thermo Scientific Chemicals

3-Chloro-2,5,6-trifluoropyridine, 98+%

CAS: 2879-42-7 | C5HClF3N | 167.52 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 L19543.14
亦稱為 L19543-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS2879-42-7
IUPAC Name3-chloro-2,5,6-trifluoropyridine
Molecular FormulaC5HClF3N
InChI KeyOROKHXKJSDPRET-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=C(F)N=C1F
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.4545-1.4585 @ 20?C
FormLiquid
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R.E. Banks.; A. Prakash.; N.D. Venayak. Studies in azide chemistry. Part IX [1]. Investigations involving fluorinated azidopyrimidines and 4-azido-3-chloro-2,5,6-trifluoropyridine. Journal of Fluorine Chemistry. 1980, 16 (4), 325-338.
  2. Carla Bobbio.; Thierry Rausis.; Manfred Schlosser Prof. Removal of Fluorine from and Introduction of Fluorine into Polyhalopyridines: An Exercise in Nucleophilic Hetarenic Substitution. Chemistry-A European Journal. 2005, 11 (6), 1903-1910.