Benzyl alcohol on polystyrene, 3.5 mmol/g, Thermo Scientific Chemicals
Benzyl alcohol on polystyrene, 3.5 mmol/g, Thermo Scientific Chemicals
Benzyl alcohol on polystyrene, 3.5 mmol/g, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl alcohol on polystyrene, 3.5 mmol/g, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19592.06
also known as L19592-06
5 g
Catalog number L19592.06
also known as L19592-06
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzyl alcohol on polystyrene
Concentration or Composition (by Analyte or Components)3.5mmol/g
MDL NumberMFCD03792087
Recommended StorageAmbient temperatures
SynonymPoly(4-hydroxymethyl)styrene
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only