3-Cyanobenzeneboronic acid, 98%
3-Cyanobenzeneboronic acid, 98%
3-Cyanobenzeneboronic acid, 98%
Thermo Scientific Chemicals

3-Cyanobenzeneboronic acid, 98%

CAS: 150255-96-2 | C7H6BNO2 | 146.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19635.03
also known as L19635-03
Price (EUR)
52,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
52,60
Each
Chemical Identifiers
CAS150255-96-2
IUPAC Name(3-cyanophenyl)boronic acid
Molecular FormulaC7H6BNO2
InChI KeyXDBHWPLGGBLUHH-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only