4-(Boc-amino)benzeneboronic acid pinacol ester, 97%
4-(Boc-amino)benzeneboronic acid pinacol ester, 97%
4-(Boc-amino)benzeneboronic acid pinacol ester, 97%
4-(Boc-amino)benzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

4-(Boc-amino)benzeneboronic acid pinacol ester, 97%

CAS: 330793-01-6 | C17H26BNO4 | 319.21 g/mol
Have Questions?
Catalog NumberQuantity
L19653.06
also known as L19653-06
5 g
Catalog number L19653.06
also known as L19653-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS330793-01-6
IUPAC Nametert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular FormulaC17H26BNO4
InChI KeyHSJNIOYPTSKQBD-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only