(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L19685.04
also known as L19685-04
Price (EUR)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS55700-44-2
IUPAC Name(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine
Molecular FormulaC26H28FeNP
InChI KeyRCAFPSZHKFOLEE-KVEBOLLKNA-N
SMILES[Fe].c1cccc1.C[C@@H](N(C)C)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)137.0-147.0?C
Optical Rotation-359.5 ± 0.1? (C=0.6 in ethanol)
FormCrystals or powder or crystalline powder
Appearance (Color)White to orange to red to brown
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine acts as a ligand used in the enantioselective vinylation of aldehydes.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Moschetta, E. G.; Gans, K. M.; Rioux, R. M. Elucidating the roles of enthalpy, entropy, and donor atom in the chelate effect for binding different bidentate ligands on the same metal center. J. Catal. 2014, 309, 11-20.
  2. Li, W. N.; Wang, Z. L. Kumada-Tamao-Corriu cross-coupling reaction of O-based electrophiles with Grignard reagents via C-O bond activation. RSC Adv. 2013, 3 (48), 25565-25575.