(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%
(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%
(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%
Thermo Scientific Chemicals

(S)-(-)-3-(Boc-amino)pyrrolidine, 99%, ee 99%

CAS: 122536-76-9 | C9H18N2O2 | 186.255 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 L19693.06
亦稱為 L19693-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS122536-76-9
IUPAC Nametert-butyl N-(pyrrolidin-3-yl)carbamate
Molecular FormulaC9H18N2O2
InChI KeyDQQJBEAXSOOCPG-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC1CCNC1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Optical Rotation-23.2? ± 0.3? (c=1 in methanol)
Enantiomeric excess≥98.5%
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only