3-Amino-3-(3-chlorophenyl)propionic acid, 98%
3-Amino-3-(3-chlorophenyl)propionic acid, 98%
3-Amino-3-(3-chlorophenyl)propionic acid, 98%
3-Amino-3-(3-chlorophenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-Amino-3-(3-chlorophenyl)propionic acid, 98%

CAS: 68208-21-9 | C9H10ClNO2 | 199.634 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19797.MD
also known as L19797-MD
Price (EUR)
44,40
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
44,40
Each
Add to cart
Chemical Identifiers
CAS68208-21-9
IUPAC Name(3R)-3-azaniumyl-3-(3-chlorophenyl)propanoate
Molecular FormulaC9H10ClNO2
InChI KeyLIDRHPCWOYOBIZ-MRVPVSSYSA-N
SMILES[NH3+][C@H](CC([O-])=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only