Furan-3-boronic acid, 97% - FAQs
Furan-3-boronic acid, 97% - FAQs
View additional product information for Furan-3-boronic acid, 97% - FAQs (L19834.03, L19834.06)
1 product FAQs found
How should I store Furan-3-boronic acid, 97%, Thermo Scientific Chemicals (Cat. No. L19834.06, L19834-06)?
We recommend storing Furan-3-boronic acid, 97%, Thermo Scientific Chemicals (Cat. No. L19834.06, L19834-06) in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.